Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGATCCTGGCATTTTCTACAGCG[A/G]CAGCTTCGGGGGCGACGCCCAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 141250 MIM: 602696 | ||||||||||||||||||||
Literature Links: |
HMOX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMOX1 - heme oxygenase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCM5 - minichromosome maintenance complex component 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006739.3 | 187 | Missense Mutation | GAC,GGC | D,G 13 | NP_006730.2 | |
XM_006724242.3 | 187 | Missense Mutation | GAC,GGC | D,G 13 | XP_006724305.1 |