Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGTCCTGGGCACAGAAGTTGTA[A/G]ACGCGACGCGTTGTCTTCACCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610877 MIM: 603560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP6R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP6R2 - protein phosphatase 6 regulatory subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SBF1 - SET binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002972.3 | 5835 | Silent Mutation | NP_002963.2 | |||
XM_005261935.2 | 5835 | Silent Mutation | XP_005261992.1 | |||
XM_011530709.2 | 5835 | Intron | XP_011529011.1 | |||
XM_011530710.2 | 5835 | Intron | XP_011529012.1 | |||
XM_017028905.1 | 5835 | Intron | XP_016884394.1 |