Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGACATCTGGGGAAGGCTCTGGC[C/T]GAGGCCCAGGTCCCAGCTCAGCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611417 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGSM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGSM1 - small G protein signaling modulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039948.3 | Intron | NP_001035037.1 | ||||
NM_001098497.2 | Intron | NP_001091967.1 | ||||
NM_001098498.2 | Intron | NP_001091968.1 | ||||
NM_133454.3 | Intron | NP_597711.1 |