Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGTGGCTGGTGCAACTCGGTTAG[C/T]AGGTGGCAGAGTGAGGATGGGACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CABP7 - calcium binding protein 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMAT5 - zinc finger matrin-type 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003692.1 | Intron | NP_001003692.1 | ||||
NM_001318129.1 | Intron | NP_001305058.1 | ||||
NM_019103.2 | Intron | NP_061976.1 |