Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTTCCCATGATGCTTATTTTTA[C/T]CTGGGCTATCAGGAACAGATGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC38 - tetratricopeptide repeat domain 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017931.2 | 538 | Silent Mutation | TAC,TAT | Y,Y 154 | NP_060401.2 | |
XM_011530259.1 | 538 | Silent Mutation | TAC,TAT | Y,Y 89 | XP_011528561.1 | |
XM_011530260.2 | 538 | Silent Mutation | TAC,TAT | Y,Y 154 | XP_011528562.1 |