Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTCAGTCAATGTCACTGAGGTC[A/G]CTGGTCGGCTCCCCGTGGACGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604589 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRD1 - bromodomain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304808.1 | 2627 | Silent Mutation | NP_001291737.1 | |||
NM_001304809.1 | 2627 | Silent Mutation | NP_001291738.1 | |||
XM_011530073.1 | 2627 | Silent Mutation | XP_011528375.1 | |||
XM_017028714.1 | 2627 | Silent Mutation | XP_016884203.1 | |||
XM_017028715.1 | 2627 | Silent Mutation | XP_016884204.1 | |||
XM_017028716.1 | 2627 | Silent Mutation | XP_016884205.1 | |||
XM_017028717.1 | 2627 | Silent Mutation | XP_016884206.1 | |||
XM_017028718.1 | 2627 | Silent Mutation | XP_016884207.1 | |||
XM_017028719.1 | 2627 | Intron | XP_016884208.1 | |||
XM_017028720.1 | 2627 | Intron | XP_016884209.1 | |||
XM_017028721.1 | 2627 | Intron | XP_016884210.1 | |||
XM_017028722.1 | 2627 | Silent Mutation | XP_016884211.1 | |||
XM_017028723.1 | 2627 | Silent Mutation | XP_016884212.1 |