Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCATCTTCCGGGCCCACAAGAA[C/T]GTCCTAGCCGCCAGCAGCATCTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607712 | ||||||||||||||||||||
Literature Links: |
HIC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIC2 - hypermethylated in cancer 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015094.2 | 319 | Silent Mutation | AAC,AAT | N,N 62 | NP_055909.2 | |
XM_011530007.2 | 319 | Silent Mutation | AAC,AAT | N,N 62 | XP_011528309.1 | |
XM_011530008.2 | 319 | Silent Mutation | AAC,AAT | N,N 62 | XP_011528310.1 | |
XM_011530009.2 | 319 | Silent Mutation | AAC,AAT | N,N 62 | XP_011528311.1 | |
XM_011530010.2 | 319 | Silent Mutation | AAC,AAT | N,N 62 | XP_011528312.1 | |
XM_017028669.1 | 319 | Silent Mutation | AAC,AAT | N,N 44 | XP_016884158.1 |