Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCCCGCAACGGTCATCTTTCC[A/G]GATAAAGCTGGCCTTCAGGAACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUSBP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUSBP11 - glucuronidase, beta pseudogene 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGL4 - ral guanine nucleotide dissociation stimulator like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153615.1 | 1430 | Missense Mutation | CAG,CGG | Q,R 87 | NP_705843.1 | |
XM_017028767.1 | 1430 | Intron | XP_016884256.1 |