Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTATTTGTGAAGAAATGGATCAG[A/G]CAACTGGAGAACCACAGTGTGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC117 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC117 - coiled-coil domain containing 117 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284263.1 | 582 | Missense Mutation | ACA,GCA | T,A 118 | NP_001271192.1 | |
NM_001284264.1 | 582 | Intron | NP_001271193.1 | |||
NM_001284265.1 | 582 | Missense Mutation | ACA,GCA | T,A 4 | NP_001271194.1 | |
NM_173510.3 | 582 | Missense Mutation | ACA,GCA | T,A 136 | NP_775781.1 |