Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCCTCGCGGAAGGTGACGGTGA[C/T]GGAGTTGGCGGTGATGTCGGTCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608457 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBX7 - chromobox 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175709.3 | 721 | Missense Mutation | ATC,GTC | I,V 230 | NP_783640.1 | |
XM_005261413.4 | 721 | Missense Mutation | ATC,GTC | I,V 137 | XP_005261470.1 | |
XM_006724174.3 | 721 | Missense Mutation | ATC,GTC | I,V 238 | XP_006724237.1 | |
XM_006724175.3 | 721 | Missense Mutation | ATC,GTC | I,V 237 | XP_006724238.1 | |
XM_006724176.3 | 721 | Missense Mutation | ATC,GTC | I,V 179 | XP_006724239.1 | |
XM_006724177.3 | 721 | Missense Mutation | ATC,GTC | I,V 171 | XP_006724240.1 | |
XM_006724178.3 | 721 | Missense Mutation | ATC,GTC | I,V 145 | XP_006724241.1 | |
XM_011530025.2 | 721 | Missense Mutation | ATC,GTC | I,V 144 | XP_011528327.1 | |
XM_017028679.1 | 721 | Missense Mutation | ATC,GTC | I,V 229 | XP_016884168.1 |