Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTGCTGCTGCCATGAGGCACC[C/T]TGGGGCCTTCCTCTTCCTTCTGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605819 MIM: 613441 | ||||||||||||||||||||
Literature Links: |
PES1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PES1 - pescadillo ribosomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243225.1 | 260 | Intron | NP_001230154.1 | |||
NM_001282327.1 | 260 | Intron | NP_001269256.1 | |||
NM_001282328.1 | 260 | Intron | NP_001269257.1 | |||
NM_014303.3 | 260 | Intron | NP_055118.1 | |||
XM_006724173.1 | 260 | Intron | XP_006724236.1 | |||
XM_017028678.1 | 260 | Intron | XP_016884167.1 |
TCN2 - transcobalamin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000355.3 | 260 | Missense Mutation | CCT,CTT | P,L 4 | NP_000346.2 | |
NM_001184726.1 | 260 | Missense Mutation | CCT,CTT | P,L 4 | NP_001171655.1 |