Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCTTACCCTCAGCAGCCTGTCAC[C/T]GTGGGGTCCAGGCCTGAGCGCTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606448 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNRD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TXNRD2 - thioredoxin reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282512.1 | 1743 | Intron | NP_001269441.1 | |||
NM_006440.4 | 1743 | Silent Mutation | ACA,ACG | T,T 518 | NP_006431.2 |