Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCGGCTGTAGGTGTTGTCGAAG[C/T]GCAGGACATCTGCAGTGATGGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDC4P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDC4P - syndecan 4 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEC14L4 - SEC14 like lipid binding 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161368.1 | 993 | Intron | NP_001154840.1 | |||
NM_174977.3 | 993 | Missense Mutation | CAC,CGC | H,R 364 | NP_777637.1 | |
XM_006724235.2 | 993 | Missense Mutation | CAC,CGC | H,R 310 | XP_006724298.1 | |
XM_011530164.2 | 993 | Missense Mutation | CAC,CGC | H,R 349 | XP_011528466.1 | |
XM_017028787.1 | 993 | Missense Mutation | CAC,CGC | H,R 423 | XP_016884276.1 | |
XM_017028788.1 | 993 | Missense Mutation | CAC,CGC | H,R 395 | XP_016884277.1 | |
XM_017028789.1 | 993 | Missense Mutation | CAC,CGC | H,R 310 | XP_016884278.1 | |
XM_017028790.1 | 993 | Intron | XP_016884279.1 | |||
XM_017028791.1 | 993 | Missense Mutation | CAC,CGC | H,R 310 | XP_016884280.1 |