Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCGTCGAGGGCGCAAAGCTGCAC[G/T]CCATCCTGGGCAAGCTGGGCCCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603721 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC116 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC116 - coiled-coil domain containing 116 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2L3 - ubiquitin conjugating enzyme E2 L3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YDJC - YdjC homolog (bacterial) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017964.1 | 907 | Silent Mutation | NP_001017964.1 | |||
XM_011529906.2 | 907 | UTR 3 | XP_011528208.1 |