Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTGGGATTAAAGTACAGTTTTC[C/T]GCTGAGGAAACCTAAGGTCTTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0930 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0930 - KIAA0930 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009880.1 | 4083 | UTR 3 | NP_001009880.1 | |||
NM_015264.1 | 4083 | UTR 3 | NP_056079.1 | |||
XM_011530017.2 | 4083 | UTR 3 | XP_011528319.1 | |||
XM_017028672.1 | 4083 | UTR 3 | XP_016884161.1 |
MIR1249 - microRNA 1249 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP50 - nucleoporin 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |