Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTTGTCCAGCGGGAACTGCACG[C/T]TCAGTGTGCCCGCCAAAAACTCATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612842 | ||||||||||||||||||||
Literature Links: |
RASD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RASD2 - RASD family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014310.3 | 1157 | Missense Mutation | CTC,TTC | L,F 12 | NP_055125.2 | |
XM_005261442.4 | 1157 | Missense Mutation | CTC,TTC | L,F 12 | XP_005261499.1 | |
XM_011530040.2 | 1157 | Missense Mutation | CTC,TTC | L,F 12 | XP_011528342.1 | |
XM_017028701.1 | 1157 | Missense Mutation | CTC,TTC | L,F 24 | XP_016884190.1 | |
XM_017028702.1 | 1157 | Missense Mutation | CTC,TTC | L,F 12 | XP_016884191.1 |