Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGCCCATGCCCGGGCCTCAGCT[C/G]AGTCCACTGCTCGTCTGCCGGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615870 | ||||||||||||||||||||
Literature Links: |
IFT27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFT27 - intraflagellar transport 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177701.2 | 834 | Nonsense Mutation | TCA,TGA | S,* 135 | NP_001171172.1 | |
NM_006860.4 | 834 | Nonsense Mutation | TCA,TGA | S,* 134 | NP_006851.1 | |
XM_006724106.2 | 834 | Nonsense Mutation | TCA,TGA | S,* 135 | XP_006724169.1 | |
XM_017028540.1 | 834 | Nonsense Mutation | TCA,TGA | S,* 94 | XP_016884029.1 |
LOC105373021 - uncharacterized LOC105373021 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |