Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCAGGGCGTGCCGGCCACCTC[A/G]TAGGTCTTGTGAAAACGTGAGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNJ4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNJ4 - potassium voltage-gated channel subfamily J member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004981.1 | 1117 | Silent Mutation | TAC,TAT | Y,Y 340 | NP_004972.1 | |
NM_152868.2 | 1117 | Silent Mutation | TAC,TAT | Y,Y 340 | NP_690607.1 |
LOC101927183 - uncharacterized LOC101927183 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |