Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAGCAACCACGGTGGCAGCTCC[A/G]GGAGCACCCATCGCTCACCCCGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606084 MIM: 150571 | ||||||||||||||||||||
Literature Links: |
CDC42EP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC42EP1 - CDC42 effector protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152243.2 | 632 | Missense Mutation | AGG,GGG | R,G 72 | NP_689449.1 | |
XM_006724107.1 | 632 | Missense Mutation | AGG,GGG | R,G 72 | XP_006724170.1 | |
XM_006724108.1 | 632 | Missense Mutation | AGG,GGG | R,G 72 | XP_006724171.1 | |
XM_006724109.1 | 632 | Missense Mutation | AGG,GGG | R,G 72 | XP_006724172.1 | |
XM_017028541.1 | 632 | Missense Mutation | AGG,GGG | R,G 72 | XP_016884030.1 |
LGALS2 - galectin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |