Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAAGGGGCCTGTTTCCAGAGAGC[C/T]CTTATGGAGAACGCCATTTGGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HORMAD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HORMAD2 - HORMA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011529914.2 | 510 | Missense Mutation | CCT,TCT | P,S 57 | XP_011528216.1 | |
XM_011529917.2 | 510 | Missense Mutation | CCT,TCT | P,S 57 | XP_011528219.1 | |
XM_011529918.1 | 510 | Missense Mutation | CCT,TCT | P,S 57 | XP_011528220.1 | |
XM_017028621.1 | 510 | Missense Mutation | CCT,TCT | P,S 57 | XP_016884110.1 | |
XM_017028622.1 | 510 | Missense Mutation | CCT,TCT | P,S 57 | XP_016884111.1 | |
XM_017028624.1 | 510 | UTR 5 | XP_016884113.1 | |||
XM_017028625.1 | 510 | UTR 5 | XP_016884114.1 | |||
XM_017028626.1 | 510 | Intron | XP_016884115.1 |