Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCACAGGTCAGAAGTGGTGGG[A/T]AGCAGGCCCAGTGGATACCTTTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604414 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C22orf23 - chromosome 22 open reading frame 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207062.1 | 1156 | UTR 3 | NP_001193991.1 | |||
NM_032561.4 | 1156 | UTR 3 | NP_115950.3 | |||
XM_005261781.1 | 1156 | UTR 3 | XP_005261838.1 | |||
XM_005261782.3 | 1156 | UTR 3 | XP_005261839.1 | |||
XM_005261783.2 | 1156 | UTR 3 | XP_005261840.1 | |||
XM_005261784.2 | 1156 | UTR 3 | XP_005261841.1 |
MICALL1 - MICAL like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2F - RNA polymerase II subunit F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |