Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCACTGCTGGCTACACCCCCAC[C/T]ACTTGCTGCCCCGCCCCCCTTCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607396 | ||||||||||||||||||||
Literature Links: |
TOB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TOB2 - transducer of ERBB2, 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016272.3 | 1822 | Missense Mutation | NP_057356.1 | |||
XM_005261315.2 | 1822 | Missense Mutation | XP_005261372.1 | |||
XM_006724105.3 | 1822 | Missense Mutation | XP_006724168.1 | |||
XM_017028539.1 | 1822 | Missense Mutation | XP_016884028.1 |