Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCAATGACATGGTCTCCATGAG[C/T]GTCCCTGACTGCCTCAGCATCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICALL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MICALL1 - MICAL like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033386.3 | 355 | Silent Mutation | AGC,AGT | S,S 89 | NP_203744.1 | |
XM_005261791.3 | 355 | Missense Mutation | AGC,AGT | S,S 89 | XP_005261848.1 | |
XM_005261792.3 | 355 | Missense Mutation | AGC,AGT | S,S 89 | XP_005261849.1 | |
XM_011530471.2 | 355 | Missense Mutation | AGC,AGT | S,S 89 | XP_011528773.1 | |
XM_011530472.2 | 355 | Missense Mutation | AGC,AGT | S,S 89 | XP_011528774.1 | |
XM_011530473.2 | 355 | Missense Mutation | AGC,AGT | S,S 89 | XP_011528775.1 | |
XM_011530474.2 | 355 | Missense Mutation | AGC,AGT | S,S 5 | XP_011528776.1 | |
XM_011530475.2 | 355 | UTR 5 | XP_011528777.1 | |||
XM_011530476.2 | 355 | Missense Mutation | AGC,AGT | S,S 89 | XP_011528778.1 | |
XM_017029026.1 | 355 | Intron | XP_016884515.1 |