Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGGACTTTCTCCTGGGGAACCC[A/G]TTCAGCTCTCCAGTGGGACAGCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604702 MIM: 604700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGXB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGXB4 - HMG-box containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOM1 - target of myb1 membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135729.1 | 149 | Intron | NP_001129201.1 | |||
NM_001135730.1 | 149 | Silent Mutation | CCA,CCG | P,P 8 | NP_001129202.1 | |
NM_001135732.1 | 149 | Silent Mutation | CCA,CCG | P,P 8 | NP_001129204.1 | |
NM_005488.2 | 149 | Silent Mutation | CCA,CCG | P,P 8 | NP_005479.1 | |
XM_011529818.2 | 149 | UTR 5 | XP_011528120.1 | |||
XM_011529820.2 | 149 | UTR 5 | XP_011528122.1 | |||
XM_017028529.1 | 149 | UTR 5 | XP_016884018.1 | |||
XM_017028530.1 | 149 | UTR 5 | XP_016884019.1 |