Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGTCTCTCTCCCCTTTTAGGTG[G/T]TTTTTCTCCTCTGTGCCTATGTTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608469 | ||||||||||||||||||||
Literature Links: |
DDX17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX17 - DEAD-box helicase 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KDELR3 - KDEL endoplasmic reticulum protein retention receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006855.3 | 368 | Missense Mutation | GTT,TTT | V,F 66 | NP_006846.1 | |
NM_016657.2 | 368 | Missense Mutation | GTT,TTT | V,F 66 | NP_057839.1 |