Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCTGGGGCACGTGACCTCATGC[A/G]GCCAGTGTCCTCTGCTAGATGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506679 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100506679 - uncharacterized LOC100506679 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTLL1 - tubulin tyrosine ligase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012263.4 | Intron | NP_036395.1 | ||||
XM_006724225.3 | Intron | XP_006724288.1 | ||||
XM_011530108.2 | Intron | XP_011528410.1 | ||||
XM_011530110.2 | Intron | XP_011528412.1 | ||||
XM_017028751.1 | Intron | XP_016884240.1 | ||||
XM_017028752.1 | Intron | XP_016884241.1 | ||||
XM_017028753.1 | Intron | XP_016884242.1 |