Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAATGGCTCCACGCCCACGAGAG[G/T]CTACTTTGAAAACAAGGTGCCACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610976 | ||||||||||||||||||||
Literature Links: |
APOBEC3H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOBEC3H - apolipoprotein B mRNA editing enzyme catalytic subunit 3H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166002.2 | 281 | Missense Mutation | GGC,GTC | G,V 45 | NP_001159474.2 | |
NM_001166003.2 | 281 | Missense Mutation | GGC,GTC | G,V 45 | NP_001159475.2 | |
NM_001166004.2 | 281 | Missense Mutation | GGC,GTC | G,V 45 | NP_001159476.2 | |
NM_181773.4 | 281 | Missense Mutation | GGC,GTC | G,V 45 | NP_861438.3 | |
XM_011529990.2 | 281 | Missense Mutation | GGC,GTC | G,V 45 | XP_011528292.1 | |
XM_011529991.2 | 281 | Missense Mutation | GGC,GTC | G,V 45 | XP_011528293.1 | |
XM_011529992.2 | 281 | Missense Mutation | GGC,GTC | G,V 45 | XP_011528294.1 |