Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCAAACTTCTCATTCATGGAGAC[A/G]GTGAAGATGTCCTTCCACAGCCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608359 | ||||||||||||||||||||
Literature Links: |
LOC105373057 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105373057 - DNA-directed RNA polymerase II subunit RPB1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULT4A1 - sulfotransferase family 4A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014351.3 | 495 | Silent Mutation | ACC,ACT | T,T 258 | NP_055166.1 | |
XM_011530120.2 | 495 | Intron | XP_011528422.1 | |||
XM_011530121.1 | 495 | Silent Mutation | ACC,ACT | T,T 145 | XP_011528423.1 |