Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACCCTGGATCCCCAGATCTCCA[A/G]TCTAGGAGTGGTGGCAGTCAATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605968 MIM: 605972 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFPL1 - ret finger protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021026.2 | Intron | NP_066306.2 | ||||
XM_011530298.2 | Intron | XP_011528600.1 | ||||
XM_011530299.2 | Intron | XP_011528601.1 | ||||
XM_017028900.1 | Intron | XP_016884389.1 | ||||
XM_017028901.1 | Intron | XP_016884390.1 | ||||
XM_017028902.1 | Intron | XP_016884391.1 |
RFPL1S - RFPL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |