Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTGATGACACTGGGTCTTGTG[C/T]TCCATTTTCTCCTTGAAAGTTAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF280B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF280B - zinc finger protein 280B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080764.3 | 1840 | Silent Mutation | GAA,GAG | E,E 477 | NP_542942.2 | |
XM_011529893.2 | 1840 | Silent Mutation | GAA,GAG | E,E 477 | XP_011528195.1 | |
XM_011529895.2 | 1840 | Silent Mutation | GAA,GAG | E,E 477 | XP_011528197.1 | |
XM_011529896.2 | 1840 | Silent Mutation | GAA,GAG | E,E 477 | XP_011528198.1 | |
XM_011529897.2 | 1840 | Silent Mutation | GAA,GAG | E,E 477 | XP_011528199.1 |