Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTCACCTCATATATCACAGACC[A/C]GGTGAAGTTCAGCTGGCCAAGGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104170 MIM: 610981 | ||||||||||||||||||||
Literature Links: |
NAGA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAGA - alpha-N-acetylgalactosaminidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000262.2 | 1629 | Missense Mutation | GGG,TGG | G,W 362 | NP_000253.1 | |
XM_005261615.4 | 1629 | Missense Mutation | GGG,TGG | G,W 362 | XP_005261672.1 | |
XM_005261616.4 | 1629 | Missense Mutation | GGG,TGG | G,W 362 | XP_005261673.1 |
WBP2NL - WBP2 N-terminal like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |