Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGAGTCTCCTAACGTGTTTTCTC[C/T]CTCTTCGTTGGGAGGGAAATCAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194355 MIM: 612062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XBP1 - X-box binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNRF3 - zinc and ring finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206998.1 | Intron | NP_001193927.1 | ||||
NM_032173.3 | Intron | NP_115549.2 | ||||
XM_011530435.2 | Intron | XP_011528737.1 | ||||
XM_011530436.2 | Intron | XP_011528738.1 | ||||
XM_011530437.1 | Intron | XP_011528739.1 | ||||
XM_011530438.2 | Intron | XP_011528740.1 | ||||
XM_017028990.1 | Intron | XP_016884479.1 |