Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATTCTTTGTGCACATCAGGAT[G/T]TAGAGGGGCATGGAGTAAATCCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC117 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC117 - coiled-coil domain containing 117 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284263.1 | 498 | Missense Mutation | GTA,TTA | V,L 90 | NP_001271192.1 | |
NM_001284264.1 | 498 | Intron | NP_001271193.1 | |||
NM_001284265.1 | 498 | UTR 5 | NP_001271194.1 | |||
NM_173510.3 | 498 | Missense Mutation | GTA,TTA | V,L 108 | NP_775781.1 |