Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGTGTCGGACCGCATCAGCTC[C/T]CTGGGCAGCCAGGCCATGCAGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151410 MIM: 605663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCR - BCR, RhoGEF and GTPase activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004327.3 | 1202 | Silent Mutation | TCC,TCT | S,S 202 | NP_004318.3 | |
NM_021574.2 | 1202 | Silent Mutation | TCC,TCT | S,S 202 | NP_067585.2 |
RSPH14 - radial spoke head 14 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |