Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAGCCGGCGCCCGCTCTCCACC[A/G]TGACATAGTTGCGCTGGCAGATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603582 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPM1F - protein phosphatase, Mg2+/Mn2+ dependent 1F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOP3B - topoisomerase (DNA) III beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282112.1 | 1836 | Missense Mutation | ACG,ATG | T,M 528 | NP_001269041.1 | |
NM_001282113.1 | 1836 | Missense Mutation | ACG,ATG | T,M 528 | NP_001269042.1 | |
NM_003935.4 | 1836 | Missense Mutation | ACG,ATG | T,M 528 | NP_003926.1 | |
XM_005261811.1 | 1836 | Missense Mutation | ACG,ATG | T,M 528 | XP_005261868.1 | |
XM_005261813.2 | 1836 | Missense Mutation | ACG,ATG | T,M 392 | XP_005261870.1 | |
XM_006724349.1 | 1836 | Missense Mutation | ACG,ATG | T,M 528 | XP_006724412.1 | |
XM_006724350.1 | 1836 | Missense Mutation | ACG,ATG | T,M 528 | XP_006724413.1 | |
XM_011530482.1 | 1836 | Missense Mutation | ACG,ATG | T,M 528 | XP_011528784.1 | |
XM_017029038.1 | 1836 | Missense Mutation | ACG,ATG | T,M 528 | XP_016884527.1 | |
XM_017029039.1 | 1836 | Missense Mutation | ACG,ATG | T,M 392 | XP_016884528.1 | |
XM_017029040.1 | 1836 | Missense Mutation | ACG,ATG | T,M 392 | XP_016884529.1 | |
XM_017029041.1 | 1836 | Missense Mutation | ACG,ATG | T,M 392 | XP_016884530.1 |