Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCAGTCTGGGATTTCACCATTC[A/G]GGACGGCGATGTTGAGGGTGTCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608955 | ||||||||||||||||||||
Literature Links: |
LOC100506679 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100506679 - uncharacterized LOC100506679 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTLL1 - tubulin tyrosine ligase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012263.4 | 1338 | Missense Mutation | CCG,CTG | P,L 357 | NP_036395.1 | |
XM_006724225.3 | 1338 | Missense Mutation | CCG,CTG | P,L 260 | XP_006724288.1 | |
XM_011530108.2 | 1338 | Missense Mutation | CCG,CTG | P,L 319 | XP_011528410.1 | |
XM_011530110.2 | 1338 | Missense Mutation | CCG,CTG | P,L 243 | XP_011528412.1 | |
XM_017028751.1 | 1338 | Missense Mutation | CCG,CTG | P,L 357 | XP_016884240.1 | |
XM_017028752.1 | 1338 | Missense Mutation | CCG,CTG | P,L 357 | XP_016884241.1 | |
XM_017028753.1 | 1338 | Missense Mutation | CCG,CTG | P,L 319 | XP_016884242.1 |