Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTTCAAGGTGTTTGACCGTGAC[A/G]GCAATGGCTTCATCTCCAAGCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CABP7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CABP7 - calcium binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182527.2 | 480 | Missense Mutation | AGC,GGC | S,G 49 | NP_872333.1 | |
XM_006724161.3 | 480 | Missense Mutation | AGC,GGC | S,G 49 | XP_006724224.1 |
ZMAT5 - zinc finger matrin-type 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |