Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATGGTGACCTGGAGCAGCTGCT[C/T]GCCCTGTATCAGTCTCAGGAGGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616727 | ||||||||||||||||||||
Literature Links: |
PHF21B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHF21B - PHD finger protein 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135862.2 | 1625 | Missense Mutation | AAG,GAG | K,E 450 | NP_001129334.1 | |
NM_001242450.1 | 1625 | Missense Mutation | AAG,GAG | K,E 438 | NP_001229379.1 | |
NM_001284296.1 | 1625 | Missense Mutation | AAG,GAG | K,E 288 | NP_001271225.1 | |
NM_138415.4 | 1625 | Missense Mutation | AAG,GAG | K,E 492 | NP_612424.1 | |
XM_006724122.1 | 1625 | Missense Mutation | AAG,GAG | K,E 450 | XP_006724185.1 | |
XM_011529855.1 | 1625 | Intron | XP_011528157.1 |