Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCGGGCCCATCCTGAACTTCCT[C/G]CGGCATGGCAAGCTGGTGCTGGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616386 | ||||||||||||||||||||
Literature Links: |
KCTD17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD17 - potassium channel tetramerization domain containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282684.1 | 291 | Silent Mutation | CTC,CTG | L,L 92 | NP_001269613.1 | |
NM_001282685.1 | 291 | Silent Mutation | CTC,CTG | L,L 92 | NP_001269614.1 | |
NM_001282686.1 | 291 | Silent Mutation | CTC,CTG | L,L 92 | NP_001269615.1 | |
NM_024681.3 | 291 | Silent Mutation | CTC,CTG | L,L 92 | NP_078957.2 | |
XM_005261741.3 | 291 | Silent Mutation | CTC,CTG | L,L 92 | XP_005261798.2 | |
XM_005261742.2 | 291 | Silent Mutation | CTC,CTG | L,L 92 | XP_005261799.2 | |
XM_005261743.2 | 291 | Silent Mutation | CTC,CTG | L,L 92 | XP_005261800.2 | |
XM_005261744.2 | 291 | Silent Mutation | CTC,CTG | L,L 92 | XP_005261801.1 | |
XM_011530374.2 | 291 | Silent Mutation | CTC,CTG | L,L 92 | XP_011528676.1 | |
XM_011530377.2 | 291 | Silent Mutation | CTC,CTG | L,L 92 | XP_011528679.1 |