Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCTTGACTGTGATGGCGCAGTC[A/G]GGGCCGCCAGAGCCCAGGGAAGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604523 MIM: 610230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CELSR1 - cadherin EGF LAG seven-pass G-type receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014246.1 | 5546 | Silent Mutation | CCC,CCT | P,P 2974 | NP_055061.1 | |
XM_006724383.3 | 5546 | Silent Mutation | CCC,CCT | P,P 2974 | XP_006724446.1 | |
XM_011530553.1 | 5546 | Intron | XP_011528855.1 | |||
XM_011530554.2 | 5546 | Silent Mutation | CCC,CCT | P,P 1805 | XP_011528856.1 | |
XM_011530555.2 | 5546 | Silent Mutation | CCC,CCT | P,P 1773 | XP_011528857.1 |
TRMU - tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |