Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTACTTCTTTCGCCGGGTGGTCCT[C/G]GAACTACTGGAGCAGGGGTTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMF2 - lipase maturation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033200.2 | 2577 | Silent Mutation | TCC,TCG | S,S 700 | NP_149977.2 | |
XM_005261955.3 | 2577 | Silent Mutation | TCC,TCG | S,S 675 | XP_005262012.1 | |
XM_006724426.3 | 2577 | Silent Mutation | TCC,TCG | S,S 645 | XP_006724489.1 | |
XM_006724427.3 | 2577 | Silent Mutation | TCC,TCG | S,S 645 | XP_006724490.1 | |
XM_017029077.1 | 2577 | Silent Mutation | TCC,TCG | S,S 662 | XP_016884566.1 |
NCAPH2 - non-SMC condensin II complex subunit H2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |