Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCAGGGGCCTGGAAACCACCTA[G/T]CTTCTTGGTGGAGCCTGGGTCTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PPM1F PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPM1F - protein phosphatase, Mg2+/Mn2+ dependent 1F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014634.3 | 1509 | Missense Mutation | AGA,AGC | R,S 454 | NP_055449.1 | |
XM_011530556.1 | 1509 | Missense Mutation | AGA,AGC | R,S 286 | XP_011528858.1 | |
XM_017029108.1 | 1509 | Intron | XP_016884597.1 |