Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCCCTCCTGCAGCACCTGCAGCC[A/G]TCGCTCTGAGGGTTCCAGCCACCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608926 MIM: 133450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMID1 - EMI domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EWSR1 - EWS RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHBDD3 - rhomboid domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006724224.3 | 876 | Missense Mutation | CGG,TGG | R,W 64 | XP_006724287.1 | |
XM_011530107.2 | 876 | Missense Mutation | CGG,TGG | R,W 192 | XP_011528409.1 | |
XM_017028749.1 | 876 | Missense Mutation | CGG,TGG | R,W 195 | XP_016884238.1 | |
XM_017028750.1 | 876 | Missense Mutation | CGG,TGG | R,W 59 | XP_016884239.1 |