Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGCCCGCCCGGCCCGAGGTGTG[A/G]CCTTCCAGGCGCACAGTGTAGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 133450 MIM: 602128 MIM: 602220 | ||||||||||||||||||||
Literature Links: |
EWSR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EWSR1 - EWS RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GAS2L1 - growth arrest specific 2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278730.1 | 434 | Missense Mutation | ACC,GCC | T,A 86 | NP_001265659.1 | |
NM_006478.4 | 434 | Missense Mutation | ACC,GCC | T,A 86 | NP_006469.2 | |
NM_152236.2 | 434 | Intron | NP_689422.1 | |||
XM_011529824.1 | 434 | Missense Mutation | ACC,GCC | T,A 86 | XP_011528126.1 | |
XM_011529825.1 | 434 | Missense Mutation | ACC,GCC | T,A 86 | XP_011528127.1 | |
XM_017028533.1 | 434 | Missense Mutation | ACC,GCC | T,A 86 | XP_016884022.1 |
RASL10A - RAS like family 10 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |