Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGTCTACCCAGTCTGCACTCC[C/T]GATGGCATCGTCTTTGACTTACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607588 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985532 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107985532 - uncharacterized LOC107985532 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIL2 - peptidylprolyl isomerase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317996.1 | 284 | Silent Mutation | CCC,CCT | P,P 56 | NP_001304925.1 | |
NM_014337.3 | 284 | Silent Mutation | CCC,CCT | P,P 56 | NP_055152.1 | |
NM_148175.2 | 284 | Silent Mutation | CCC,CCT | P,P 56 | NP_680480.1 | |
NM_148176.2 | 284 | Silent Mutation | CCC,CCT | P,P 56 | NP_680481.1 | |
XM_005261448.3 | 284 | Silent Mutation | CCC,CCT | P,P 56 | XP_005261505.1 | |
XM_011530041.2 | 284 | Silent Mutation | CCC,CCT | P,P 56 | XP_011528343.1 | |
XM_011530042.2 | 284 | Silent Mutation | CCC,CCT | P,P 56 | XP_011528344.1 | |
XM_011530043.2 | 284 | Silent Mutation | CCC,CCT | P,P 56 | XP_011528345.1 | |
XM_011530044.2 | 284 | Silent Mutation | CCC,CCT | P,P 56 | XP_011528346.1 | |
XM_011530046.2 | 284 | Silent Mutation | CCC,CCT | P,P 56 | XP_011528348.1 | |
XM_011530047.2 | 284 | Silent Mutation | CCC,CCT | P,P 56 | XP_011528349.1 | |
XM_011530049.2 | 284 | UTR 5 | XP_011528351.1 | |||
XM_011530050.2 | 284 | UTR 5 | XP_011528352.1 | |||
XM_011530051.2 | 284 | UTR 5 | XP_011528353.1 | |||
XM_017028705.1 | 284 | Intron | XP_016884194.1 | |||
XM_017028706.1 | 284 | Intron | XP_016884195.1 |