Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGAGAAAGGAAAAGAAAAGAAA[C/T]AGTCTGTGTCTTACATGATTCCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113508 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C22orf24 - chromosome 22 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302819.1 | Intron | NP_001289748.1 | ||||
NM_001302820.1 | Intron | NP_001289749.1 | ||||
NM_015372.2 | Intron | NP_056187.1 |
YWHAH - tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |