Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCTGTTGCTGCCGTGTGCCCCC[A/G]GCCCCTAGTTCTGGCGCCCCATCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CECR5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CECR5 - cat eye syndrome chromosome region, candidate 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017829.5 | 1521 | Silent Mutation | GCC,GCT | A,A 312 | NP_060299.4 | |
NM_033070.2 | 1521 | Silent Mutation | GCC,GCT | A,A 342 | NP_149061.1 | |
XM_005261236.3 | 1521 | Silent Mutation | GCC,GCT | A,A 181 | XP_005261293.1 | |
XM_011546126.2 | 1521 | Silent Mutation | GCC,GCT | A,A 274 | XP_011544428.1 | |
XM_011546127.1 | 1521 | Silent Mutation | GCC,GCT | A,A 220 | XP_011544429.1 | |
XM_017028781.1 | 1521 | Silent Mutation | GCC,GCT | A,A 181 | XP_016884270.1 | |
XM_017028782.1 | 1521 | Silent Mutation | GCC,GCT | A,A 153 | XP_016884271.1 | |
XM_017028783.1 | 1521 | Silent Mutation | GCC,GCT | A,A 153 | XP_016884272.1 | |
XM_017028784.1 | 1521 | Silent Mutation | GCC,GCT | A,A 153 | XP_016884273.1 |
LOC100996342 - uncharacterized LOC100996342 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |