Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTTCCTGGTGGTTCTCCGAGG[C/T]GTACAGGCGCCCATCGCGCACGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611746 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373051 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105373051 - uncharacterized LOC105373051 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCUBE1 - signal peptide, CUB domain and EGF like domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173050.3 | 2912 | Missense Mutation | ACC,GCC | T,A 929 | NP_766638.2 | |
XM_005261752.1 | 2912 | Missense Mutation | ACC,GCC | T,A 959 | XP_005261809.1 | |
XM_011530389.2 | 2912 | Missense Mutation | ACC,GCC | T,A 853 | XP_011528691.1 | |
XM_011530390.2 | 2912 | Missense Mutation | ACC,GCC | T,A 853 | XP_011528692.1 |