Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCAAAGCACCGCAACATGACC[A/G]AAATGAGCTTCCTGAGCAGCGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604064 MIM: 615497 MIM: 610225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATF4 - activating transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001675.4 | 766 | Intron | NP_001666.2 | |||
NM_182810.2 | 766 | Intron | NP_877962.1 | |||
XM_017028807.1 | 766 | Missense Mutation | AAA,GAA | K,E 3 | XP_016884296.1 |
MIEF1 - mitochondrial elongation factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS19BP1 - ribosomal protein S19 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |