Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCACAAATCTGCCTGCAGCTG[C/T]AGTCTTGAGTGCCCAGGTGTACAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612214 | ||||||||||||||||||||
Literature Links: |
GUSBP11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GUSBP11 - glucuronidase, beta pseudogene 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGL4 - ral guanine nucleotide dissociation stimulator like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153615.1 | 1205 | Missense Mutation | GCA,GTA | A,V 12 | NP_705843.1 | |
XM_017028767.1 | 1205 | Intron | XP_016884256.1 |